Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W269018

Sigma-Aldrich

Methyl p-tert-butylphenylacetate

≥97%, FG

Synonym(s):

Methyl 2-(4-tert-butylphenyl)acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CC6H4CH2CO2CH3
CAS Number:
Molecular Weight:
206.28
FEMA Number:
2690
EC Number:
Council of Europe no.:
577
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.758
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥97%

refractive index

n20/D 1.501 (lit.)

bp

106 °C/2 mmHg (lit.)

density

0.999 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

green; waxy; fruity

SMILES string

COC(=O)Cc1ccc(cc1)C(C)(C)C

InChI

1S/C13H18O2/c1-13(2,3)11-7-5-10(6-8-11)9-12(14)15-4/h5-8H,9H2,1-4H3

InChI key

HXVTYMWVMVKVTF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl p-tert-butylphenylacetate can be used as a food flavoring agent.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Encyclopedia of Food and Color Additives, 1, 1737-1737 (1997)
Encyclopedia of Food and Color Additives, 1, 1737-1737 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service