Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W266809

Sigma-Aldrich

DL-Menthyl acetate

≥97%, FG

Synonym(s):

(5-methyl-2-propan-2-ylcyclohexyl) acetate, Menthyl acetate racemic

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2668
EC Number:
Council of Europe no.:
206
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.016
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥97%

refractive index

n20/D 1.445

bp

228-229 °C

density

0.922 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

berry; cooling; minty

SMILES string

CC(C)C1CCC(C)CC1OC(C)=O

InChI

1S/C12H22O2/c1-8(2)11-6-5-9(3)7-12(11)14-10(4)13/h8-9,11-12H,5-7H2,1-4H3

InChI key

XHXUANMFYXWVNG-UHFFFAOYSA-N

Related Categories

Biochem/physiol Actions

Taste at 25 ppm

Other Notes

Natural occurrence: Mentha arvensis, peppermint, scotch spearmint, raspberry, mango, buchu oil, lemon blam, ginger and orange juice, eucalyptus oil.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Mosciano, G.
Perfumer & Flavorist, 27, 58-58 (2002)
Mosciano, G.
Perfumer & Flavorist, 30, 55-55 (2005)
Organoleptic Characteristics of Flavor Materials
Mosciano, G.
Perfumer & Flavorist, 28, 77-77 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service