Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

W262255

Sigma-Aldrich

Bulgarian lavender oil

greener alternative

natural, FG, product of Essential oils and Herbs Ltd

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2622
UNSPSC Code:
12164502
NACRES:
NA.21

biological source

Lavandula angustifolia

Quality Level

grade

FG
Fragrance grade
natural

Agency

follows IFRA guidelines

reg. compliance

EU 1334/2008 & 178/2002
EU Regulation 1223/2009
FDA 21 CFR 117

composition

Contains EU 1223/2009 restricted Limonene and Linalool and IFRA and EU1223/2009 restricted Geraniol and Coumarin

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

limonene (sum of D, L and DL), coumarin, linalool, geraniol

greener alternative category

Organoleptic

fresh; lavender; floral

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.
Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Eye Irrit. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

172.4 °F

Flash Point(C)

78 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Essential oils are complex natural extracts consisting of aromatic compounds and related organic chemicals. While they hold great value as fragrance and flavor ingredients, their significant risk of adulteration necessitates consistent formulator testing for quality, naturalness, and allergen levels.

Essential oils are complex natural extracts consisting of aromatic compounds and related organic chemicals. While they hold great value as fragrance and flavor ingredients, their significant risk of adulteration necessitates consistent formulator testing for quality, naturalness, and allergen levels.

Essential oils are complex natural extracts consisting of aromatic compounds and related organic chemicals. While they hold great value as fragrance and flavor ingredients, their significant risk of adulteration necessitates consistent formulator testing for quality, naturalness, and allergen levels.

Essential oils are complex natural extracts consisting of aromatic compounds and related organic chemicals. While they hold great value as fragrance and flavor ingredients, their significant risk of adulteration necessitates consistent formulator testing for quality, naturalness, and allergen levels.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service