Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W259525

Sigma-Aldrich

β-Ionone

natural, ≥95%, FG

Synonym(s):

4-(2,6,6-Trimethyl-1-cyclohexenyl)-3-buten-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H20O
CAS Number:
Molecular Weight:
192.30
FEMA Number:
2595
Beilstein:
1909544
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.008
NACRES:
NA.21
grade:
FG
Halal
Kosher
natural
food allergen:
no known allergens

grade

FG
Halal
Kosher
natural

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥95%

refractive index

n20/D 1.52 (lit.)

bp

126-128 °C/12 mmHg (lit.)

density

0.945 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

berry; floral; woody; sweet

SMILES string

CC(=O)\C=C\C1=C(C)CCCC1(C)C

InChI

1S/C13H20O/c1-10-6-5-9-13(3,4)12(10)8-7-11(2)14/h7-8H,5-6,9H2,1-4H3/b8-7+

InChI key

PSQYTAPXSHCGMF-BQYQJAHWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

β-Ionone is one of the main volatile compounds identified in green tea and tomatoes.

Other Notes

Natural occurrence: Tobacco, grape, grape brandy, orange juice, papaya, peach, raspberry, roasted almond, tea and spearmint.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enzymatic modification of tomato homogenate and its effect on volatile flavor compounds.
Yilmaz E, et al.
Journal of Food Science, 67(6), 2122-2125 (2002)
Further Investigation of Flavor Constituents in Manufactured Green Tea
Yamanishi T, et al.
Agricultural and Biological Chemistry, 34(4), 599-608 (1970)
Vishal Sharma et al.
Bioorganic & medicinal chemistry letters, 22(20), 6343-6346 (2012-09-25)
A series of chalcones (3a-v) have been synthesized by condensation of β-ionone (1) with a variety of aldehydes (2a-v). The synthesized compounds have been screened for their in vitro antimicrobial activity against five bacterial and five fungal strains, using disc
Chin-Shiu Huang et al.
The British journal of nutrition, 107(5), 631-638 (2011-07-27)
β-Ionone (BI), a precursor for carotenoids, is widely distributed in fruit and vegetables. Recent in vitro studies have demonstrated the potential anti-metastatic effects of BI, but the mechanisms underlying such actions are not clear. Because liver cancer is the most
Jihai Shao et al.
Aquatic toxicology (Amsterdam, Netherlands), 104(1-2), 48-55 (2011-05-06)
In order to explore the potential targets of toxicity of β-ionone on the photosynthetic system of Microcystis aeruginosa, the polyphasic rise in chlorophyll a (Chl a) fluorescence transient and transcript expression for key genes in photosystem II (PSII) of M.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service