Skip to Content
MilliporeSigma
All Photos(1)

Documents

W202118

Sigma-Aldrich

Allyl butyrate

≥98%, FG

Synonym(s):

Allyl butanoate, Butanoic acid 2-propen-1-yl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CO2CH2CH=CH2
CAS Number:
Molecular Weight:
128.17
FEMA Number:
2021
EC Number:
Council of Europe no.:
280
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.054
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.414 (lit.)

bp

44-45 °C/15 mmHg (lit.)

density

0.902 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; waxy; fruity; sweet

SMILES string

CCCC(=O)OCC=C

InChI

1S/C7H12O2/c1-3-5-7(8)9-6-4-2/h4H,2-3,5-6H2,1H3

InChI key

RMZIOVJHUJAAEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

107.6 °F - closed cup

Flash Point(C)

42 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service