Key Documents
TEM-LC400CU25
Sigma-Aldrich
Lacey Carbon Supported Copper Grids
size 400 mesh, size 50 nm , hole size, box of 25 ×
Synonym(s):
Lacey/Carbon Film, 400 Mesh, Copper, 25pk
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
packaging
box of 25 ×
size
400 mesh
50 nm , hole size
General description
Lacey carbon film supported TEM grids. This carbon coating consists of woven-mesh-like openings of different sizes and shapes. This type of pattern provides support but does not interfere when observing specimen sections.
Choose from one of the most recent versions:
Certificates of Analysis (COA)