Key Documents
TEM-HC300CUCC
Sigma-Aldrich
Holey Carbon/Continuous Ultrathin carbon film Supported Copper Grids
size 300 mesh, box of 25 ×
Synonym(s):
Holey Carbon Film - Cu, 300 mesh, less than 3 nm, CC
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
packaging
box of 25 ×
size
300 mesh
General description
Holey Carbon Film with a Continuous Layer of Ultrathin Carbon Film (< 3 nm thick) on 300 mesh copper grids.
Choose from one of the most recent versions:
Certificates of Analysis (COA)