Key Documents
TEM-FCF100AU
Sigma-Aldrich
Formvar/Carbon Supported Gold Grids
size 100 mesh, box of 25 ×
Synonym(s):
Formvar/Carbon Square Mesh, AU, 100 Mesh
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
packaging
box of 25 ×
size
100 mesh
General description
A formvar coated TEM grid, stabilized with evaporated carbon film. This type of coating is excellent for specimen support, especially for ultra thin sections.
Thickness: 10nm Formvar and 1nm Carbon
Choose from one of the most recent versions:
Certificates of Analysis (COA)