All Photos(1)
About This Item
Linear Formula:
(CH3O)3C6H2NH2
CAS Number:
Molecular Weight:
183.20
Beilstein:
642919
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
mp
110-113 °C (lit.)
SMILES string
COc1cc(N)cc(OC)c1OC
InChI
1S/C9H13NO3/c1-11-7-4-6(10)5-8(12-2)9(7)13-3/h4-5H,10H2,1-3H3
InChI key
XEFRNCLPPFDWAC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Romeo Romagnoli et al.
Bioorganic chemistry, 97, 103665-103665 (2020-02-23)
A new class of inhibitors of tubulin polymerization based on the 2-alkoxycarbonyl-3-(3',4',5'-trimethoxyanilino)indole molecular skeleton was synthesized and evaluated for antiproliferative activity, inhibition of tubulin polymerization and cell cycle effects. The results presented show that the methoxy substitution and location on
Gina Ambrosio et al.
Scientific reports, 10(1), 4114-4114 (2020-03-07)
Chemical reaction with diazonium molecules has revealed to be a powerful method for the surface chemical modification of graphite, carbon nanotubes and recently also of graphene. Graphene electronic structure modification using diazonium molecules is strongly influenced by graphene growth and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service