All Photos(2)
About This Item
Linear Formula:
(CH3)4C6H6(=O)
CAS Number:
Molecular Weight:
154.25
Beilstein:
1237421
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
refractive index
n20/D 1.451 (lit.)
density
0.881 g/mL at 25 °C (lit.)
SMILES string
CC1(C)CC(=O)CC(C)(C)C1
InChI
1S/C10H18O/c1-9(2)5-8(11)6-10(3,4)7-9/h5-7H2,1-4H3
InChI key
OQJMHUOCLRCSED-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
194.0 °F - closed cup
Flash Point(C)
90 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service