Key Documents
SYNM500044
Synthware vacuum manifold with PTFE stopcocks
number of ports, 4, Hose Connection: left, stopcock size 4 mm
Synonym(s):
Synthware M500044, vacuum unit
About This Item
UNSPSC Code:
40161501
NACRES:
NB.51
feature
number of ports 4
packaging
pkg of 1 ea
manufacturer/tradename
Synthware M500044
stopcock size
4 mm
General description
General purpose manifold with 2 mm or 4 mm PTFE stopcocks. Main tube is 22 mm O.D. with serrated hose connection(10 mm O.D.) at left side. Delivery ports have 10 mm serrated hose connections. Distance between stopcocks is 75 mm.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)