Key Documents
SYNF771520M
Synthware Gooch low form crucible
15 mL, frit O.D. 20 mm, porosity: medium
Synonym(s):
Synthware F771520M, Gooch filter
About This Item
UNSPSC Code:
41122200
NACRES:
NB.51
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F771520M
frit O.D.
20 mm
capacity
15 mL
Related Categories
General description
Synthware Gooch crucible with fritted disc (O.D. 20 mm) and medium porosity. Made from durable borosilicate glass.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)