Key Documents
SYNF611410
Synthware pressure equalizing addition funnel with metering stopcock
10 mL, joint: ST/NS 14/20
Synonym(s):
Synthware F611410, dropping funnel
About This Item
UNSPSC Code:
41121809
NACRES:
NB.51
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F611410
capacity
10 mL
10 mL
General description
With PTFE adjustable bore metering plug. Turning the adjustment knob allows flow rate to be controlled from full flow to drop wise. The metering plug completely solves the problems of sudden overflow or non-flow during addition of reagents.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)