Key Documents
SYNF541250
Synthware reaction flask with high vacuum valve
250 mL, joint: ST/NS 14/20, valve size 0-4 mm
Synonym(s):
Synthware F541250, Schlenk flask
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
material
borosilicate glass flask
round bottom
single-neck (high vaccum valve)
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F541250
capacity
250 mL
250 mL
valve size
0-4 mm
General description
Supplied with high vacuum valve with PTFE O-ring at tip and PTFE wiper to protect Viton® O-ring on shaft.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)