Key Documents
SYNF501010
Synthware Erlenmeyer flask
capacity 10 mL, joint: ST/NS 14/10
Synonym(s):
Synthware F501010, microscale flask
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
material
black Nylon cap
flat bottom
glass flask
Joint
joint neck14/20
joint: ST/NS14/10
sterility
non-sterile
feature
with cap
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F501010
capacity
10 mL
General description
Supplied with one compression cap, one Viton® O-ring and one PTFE-faced silicone rubber septum. For replacement caps, see RC112; for Viton® O-ring s, see R014; for septa, see R1420.
Other Notes
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)