Key Documents
SYNF451025A
Synthware Squibb-style separatory funnel with standard taper joints, PTFE stopcock and PTFE stopper
25 mL, joint: ST/NS 14/20, PTFE stopcock and glass stopper
Synonym(s):
Synthware F451025A, glass separatory funnel
About This Item
UNSPSC Code:
41121809
NACRES:
NB.51
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F451025A
capacity
25 mL
25 mL
General description
With 2 mm PTFE stopcock. Supplied with a glass stopper or PTFE stopper.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)