Key Documents
SYNF431500
Synthware three neck round bottom flask with angled side necks
500 mL, center joint: ST/NS 14/20, side joint: ST/NS 14/20
Synonym(s):
Synthware F431500, 3-neck round flask
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
material
borosilicate glass flask
round bottom
three-neck
Joint
center joint: ST/NS14/20
side joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F431500
capacity
500 mL
500 mL
General description
Round bottom with joints. Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)