Key Documents
SYNF291210
Synthware solvent addition funnel
top diam. 100 mm
Synonym(s):
Synthware F291210, glass funnel
About This Item
UNSPSC Code:
41121809
NACRES:
NB.51
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F291210
top diam.
100 mm
General description
Design of funnel permits addition of solvents to flash chromatography columns without disturbing the silica-gel/packing layer. Lower end of the funnel is closed and the stem has a series of small holes through which the solvent drains. Top of the funnel is 90mm in diameter with the stem size as listed.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)