MilliporeSigma
SYNC1301
Synthware chromatography flow control adapter
with PTFE stopcock
Synonym(s):
Synthware C1301, Glass chromatography adapter
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.51
UNSPSC Code:
41121800
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C1301
suitability
suitable for chromatography
Related Categories
General description
0-4 mm high vacuum valve with metering tip to adjust air pressure. With standard taper joint. Order CC3525 pinch type clamps with screw-locking device for ball joints. For replacement O-rings on high vacuum plug shaft, order R010. The O-rings on the high vacuum PTFE valve are completely encapsulated with thin layer of PTFE, making it inert to chemicals.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
If you need assistance, please contact Customer Support
Already Own This Product?
Visit the Document Library
Global Trade Item Number
SKUGTIN
SYNC1301-1EA04061832190235