Key Documents
SYNB262924
Synthware bubbler with PTFE stopcock
top joint: ST/NS 29/42, bottom joint: ST/NS 24/40
Synonym(s):
Synthware B262924, mineral oil bubbler
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Joint
bottom joint: ST/NS24/40
joint: ST/NS top29/42
packaging
pkg of 1 ea
manufacturer/tradename
Synthware B262924
Related Categories
General description
With small ball joints to prevent mineral oil being sucked back under sudden negative pressure in the system.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)