Key Documents
SYNA881420G
Synthware vacuum/inert gas adapter with T-bore glass stopcock
joint: ST/NS 14/20
Synonym(s):
Synthware A881420G, T-shape vacuum gas adapter
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A881420G
General description
With 4 mm T-bore glass stopcock having constant pressure retention device or 2 mm T-bore 1:5 taper PTFE plug.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)