Key Documents
SYNA201924
Synthware reducing adapter
top joint: ST/NS 19/22, bottom joint: ST/NS 24/40
Synonym(s):
Synthware A201924, glass reducing adapter, connecting adapter
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
Joint
bottom joint: ST/NS24/40
joint: ST/NS top19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A201924
General description
Synthware reducing adapter is used for connecting dissimilar standard taper joint sizes. The bottom joint (24/40) is larger than the top joint (19/22).
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)