Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SIK7122-10

Sigma-Aldrich

12,12,13,13,14,14,15,15,16,16,17,17,18,18,19,19,19-Heptadecafluorononadecylphosphonic acid

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H24F17O3P
CAS Number:
Molecular Weight:
654.34
UNSPSC Code:
12352106
NACRES:
NA.23

Assay

95%

form

powder

shelf life

3 years under the recommended conditions

color

white

Application

These products are used to form water repellent and/or lubricated surfaces on metal oxide materials (TiO2, ZnO, etc.).
The fluorinated carbon gives the omniphobic properties to the surface. The coated surface is more water repellent when a product having more fluorinated carbons is used.
The phosphonic acids groups can covalently bond to metal oxide surfaces to form sufficient self-assembled monolayer.

Legal Information

Product of SiKEMIA

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service