Skip to Content
MilliporeSigma
All Photos(1)

Documents

QBD11163

Sigma-Aldrich

DOTA-tris(acid)-amido-dPEG ®24-TFP ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C77H131F10N5O37
Molecular Weight:
1908.86
UNSPSC Code:
12352106
NACRES:
NA.22

Assay

>90%

form

solid or viscous liquid

reaction suitability

reagent type: chelator
reagent type: cross-linking reagent
reagent type: fluorescent-labelling reagent

polymer architecture

shape: linear
functionality: monofunctional

shipped in

ambient

storage temp.

−20°C

Features and Benefits

DOTA-tris(acid)-amido-dPEG ®₂₄-TFP ester contains a DOTA-tris(acid) metal chelator for radio-labelling connected through a hydrophilic dPEG spacer to a TFP ester which can react with a primary amine.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service