Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

QBD10785

Sigma-Aldrich

Biotin-dPEG®23-MAL

Synonym(s):

Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C65H119N5O28S
Molecular Weight:
1450.72
UNSPSC Code:
51171641
PubChem Substance ID:
NACRES:
NA.22

Assay

>90%

form

solid or viscous liquid

reaction suitability

reaction type: Biotinylations
reactivity: thiol reactive

reagent type: cross-linking reagent

polymer architecture

shape: linear
functionality: monofunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C(CCCCC1SCC(N2)C1NC2=O)NCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCNC(CCN3C(C=CC3=O)=O)=O

Features and Benefits

Biotin-dPEG®23-MAL takes advantage of the well-known, specific Michael reaction between a maleimide group and a free sulfhydryl to allow site-specific biotin labeling of molecules in aqueous media within the pH range of 6.5 – 7.5. The single molecular weight dPEG® spacer (94.1 Å) provide precise spacing between the biotin moiety and the labeled molecule. The dPEG® spacer imparts water solubility to the labeled molecule while reducing immunogenicity, aggregation, and precipitation of labeled biomolecules or other labeled compounds.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service