Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

QBD10093

Sigma-Aldrich

t-Boc-N-amido-dPEG®23-amine

>95% (HPLC)

Synonym(s):

t-Boc-N-amido-PEG23-amine, t-Boc-NH-PEG1000-amine, t-Boc-NH-PEG23-amine, t-Boc-PEG-amine, Boc-NH-PEG23-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C53H108N2O25
Molecular Weight:
1173.42
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.22

Assay

>95% (HPLC)

form

solid or viscous liquid

reaction suitability

reaction type: Pegylations

polymer architecture

shape: linear
functionality: homobifunctional

shipped in

ambient

storage temp.

−20°C

Features and Benefits

t-Boc-N-amido-dPEG®23-amine provides two amino groups — a tert-butyl carbamate protects one of the two amines — for reactions with aldehydes, ketones, and carboxylic acids and their active esters. The single molecular weight, discrete chain length polyethylene glycol (dPEG®) linker between the two reactive ends is 74 atoms long (86.5 Å). The tert-butyl-carbamate protecting group cleaves rapidly under mild conditions. t-Boc-N-amido-dPEG®23-amine is a monodisperse equivalent of polymer PEG1000. Because one end of this monodispersed PEG product is protected, the user has tighter control over crosslinking reactions.

Legal Information

Products Protected under U.S. Patent #s 7,888,536 & 8,637,711 and European Patent #s 1,594,440 & 2,750,681
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service