Documents
PL800CARTGB
Plas-Labs transport support cart
fits GB, NB, or AC style glove boxes
About This Item
UNSPSC Code:
41103406
NACRES:
NB.35
material
stainless steel
feature
Anti-scuff casters including brakes and locks
packaging
pkg of 1 ea
W × D × H
51 in. × 24 in. × 35 in.
General description
Suspended sliding shelf for single operator units. Culture plate capacity is about 340. Fabricated of .375 in. thick clear acrylic, with shelves spaced at 7 in. apart. The entire assembly is suspended from the chamber top to keep the floor area uncluttered.
All carts have anti-scuff casters including brakes and locks. Carts also feature a cushioning system that minimizes vibration, shocks, and wear and tear on chambers.