Skip to Content
MilliporeSigma
All Photos(1)

Documents

PCL500

Sigma-Aldrich

PCL Nanoparticles

500 nm average diameter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

average diameter

500 nm

Looking for similar products? Visit Product Comparison Guide

General description

PCL particles are made with poly(Ɛ-caprolactone) with a MW of 14,000.10 mg/ml particle suspension in DI water. Sonicate or vortex the particles before each use.

Legal Information

Product of Phosphorex, Inc.Degradex(RM) is a product of Phosphorex, Inc.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Polymeric antioxidants show promise in drug delivery with superior bioavailability and antioxidant potential.

Polymeric antioxidants show promise in drug delivery with superior bioavailability and antioxidant potential.

Polymeric antioxidants show promise in drug delivery with superior bioavailability and antioxidant potential.

Polymeric antioxidants show promise in drug delivery with superior bioavailability and antioxidant potential.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service