Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PCL10K

Sigma-Aldrich

PCL Nanoparticles

10 μm average diameter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

average diameter

10 μm

Looking for similar products? Visit Product Comparison Guide

General description

PCL particles are made with poly(Ɛ-caprolactone) with a MW of 14,000.10 mg/ml particle suspension in DI water. Sonicate or vortex the particles before each use.

Legal Information

Product of Phosphorex, Inc.Degradex(RM) is a product of Phosphorex, Inc.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Polymeric antioxidants show promise in drug delivery with superior bioavailability and antioxidant potential.

Polymeric antioxidants show promise in drug delivery with superior bioavailability and antioxidant potential.

Polymeric antioxidants show promise in drug delivery with superior bioavailability and antioxidant potential.

Polymeric antioxidants show promise in drug delivery with superior bioavailability and antioxidant potential.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service