Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M9203

Sigma-Aldrich

2′-Methoxyacetophenone

99%

Synonym(s):

2-Acetylanisole

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4COCH3
CAS Number:
Molecular Weight:
150.17
Beilstein:
1100602
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.5393 (lit.)

bp

131 °C/18 mmHg (lit.)

density

1.09 g/mL at 25 °C (lit.)

SMILES string

COc1ccccc1C(C)=O

InChI

1S/C9H10O2/c1-7(10)8-5-3-4-6-9(8)11-2/h3-6H,1-2H3

InChI key

DWPLEOPKBWNPQV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Victoria Ruiz-Hernández et al.
Frontiers in plant science, 12, 647347-647347 (2021-09-10)
Studies on the selection of floral traits usually consider pollinators and sometimes herbivores. However, humans also exert selection on floral traits of ornamental plants. We compared the preferences of bumblebees (Bombus terrestris), thrips (Frankliniella occidentalis), and humans for flowers of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service