Skip to Content
MilliporeSigma
All Photos(1)

Documents

M80202

Sigma-Aldrich

2-Methylquinoxaline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8N2
CAS Number:
Molecular Weight:
144.17
Beilstein:
113307
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.613 (lit.)

bp

245-247 °C (lit.)

density

1.118 g/mL at 25 °C (lit.)

SMILES string

Cc1cnc2ccccc2n1

InChI

1S/C9H8N2/c1-7-6-10-8-4-2-3-5-9(8)11-7/h2-6H,1H3

InChI key

ALHUXMDEZNLFTA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

224.6 °F - closed cup

Flash Point(C)

107 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Methylglyoxal assay in cells as 2-methylquinoxaline using 1,2-diaminobenzene as derivatizing reagent.
C Cordeiro et al.
Analytical biochemistry, 234(2), 221-224 (1996-02-15)
Determination of methylglyoxal as 2-methylquinoxaline by high-performance liquid chromatography and its application to biological samples.
S Ohmori et al.
Journal of chromatography, 414(1), 149-155 (1987-02-20)
F W Chaplen et al.
Proceedings of the National Academy of Sciences of the United States of America, 95(10), 5533-5538 (1998-05-20)
Methylglyoxal is an alpha-ketoaldehyde and dicarbonyl formed in cells as a side product of normal metabolism. Endogenously produced dicarbonyls, such as methylglyoxal, are involved in numerous pathogenic processes in vivo, including carcinogenesis and advanced glycation end-product formation; advanced glycation end-products
Fadhel A Alomar et al.
Antioxidants (Basel, Switzerland), 9(7) (2020-07-10)
Accumulation of methylglyoxal (MG) arising from downregulation of its primary degrading enzyme glyoxalase-1 (Glo1) is an underlying cause of diabetic cardiomyopathy (DC). This study investigated if expressing Glo1 in rat hearts shortly after the onset of Type 1 diabetes mellitus
Weerachat Sompong et al.
BMC complementary and alternative medicine, 15, 394-394 (2015-11-02)
Methylglyoxal (MG) is one of the most reactive glycating agents, which result the formation of advanced glycation end-products (AGEs) that have been implicated in the progression of age-related diseases. Inhibition of MG-induced AGE formation is the imperative approach for alleviating

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service