Skip to Content
MilliporeSigma
All Photos(1)

Documents

M23527

Sigma-Aldrich

3-(4-Methoxyphenyl)propionic acid

98%

Synonym(s):

4-Methoxyhydrocinnamic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CH2CH2CO2H
CAS Number:
Molecular Weight:
180.20
Beilstein:
1953326
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

98-100 °C (lit.)

SMILES string

COc1ccc(CCC(O)=O)cc1

InChI

1S/C10H12O3/c1-13-9-5-2-8(3-6-9)4-7-10(11)12/h2-3,5-6H,4,7H2,1H3,(H,11,12)

InChI key

FIUFLISGGHNPSM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naoki Umezawa et al.
Chembiochem : a European journal of chemical biology, 16(12), 1811-1819 (2015-07-22)
A versatile solid-phase approach based on peptide chemistry was used to construct four classes of structurally diverse polyamines with modified backbones: linear, partially constrained, branched, and cyclic. Their effects on DNA duplex stability and structure were examined. The polyamines showed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service