LGFR200
Red Fluorescent PLGA nanoparticles
200 nm average diameter
About This Item
Recommended Products
description
Excitation/Emission: 530/582 nm
Excitation/Emission: 460/500 nm
form
pellets
average diameter
200 nm
storage temp.
-10 to -25°C
Looking for similar products? Visit Product Comparison Guide
General description
- Warm to ambient temperature
- Add DI water or buffer to achieve desired concentration
- Vortex and sonicate for 5 minutes to ensure particles are well suspended
Application
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Polymeric antioxidants show promise in drug delivery with superior bioavailability and antioxidant potential.
Polymeric antioxidants show promise in drug delivery with superior bioavailability and antioxidant potential.
Polymeric antioxidants show promise in drug delivery with superior bioavailability and antioxidant potential.
Polymeric antioxidants show promise in drug delivery with superior bioavailability and antioxidant potential.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service