Skip to Content
MilliporeSigma
All Photos(1)

Documents

L754080

Sigma-Aldrich

ADT

AldrichCPR, sublimed

Synonym(s):

Anthra[2,3-b:6,7-b′]dithiophene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H10S2
CAS Number:
Molecular Weight:
290.40
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

sublimed

loss

0.5 wt. %, 280 °C

λmax

320 nm in methylene chloride

SMILES string

c1cc2cc3cc4cc5sccc5cc4cc3cc2s1

InChI

1S/C18H10S2/c1-3-19-17-9-15-8-14-6-12-2-4-20-18(12)10-16(14)7-13(15)5-11(1)17/h1-10H

InChI key

DAMUWSYTQPWFIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Anthra[2,3-b:6,7-b′]dithiophene (ADT) is popularly used as Single mol. switches in organic electronics [10]

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher J Sweeney et al.
The New England journal of medicine, 373(8), 737-746 (2015-08-06)
Androgen-deprivation therapy (ADT) has been the backbone of treatment for metastatic prostate cancer since the 1940s. We assessed whether concomitant treatment with ADT plus docetaxel would result in longer overall survival than that with ADT alone. We assigned men with
Florence K Keane et al.
Cancer, 121(16), 2713-2719 (2015-05-01)
Radiotherapy (RT), short-course androgen deprivation therapy (ADT), and brachytherapy in various combinations are treatment options for patients with intermediate-risk prostate cancer (PC), but the question of which combination if any is necessary to minimize PC-specific mortality (PCSM) risk in patients
B A Mahal et al.
Annals of oncology : official journal of the European Society for Medical Oncology, 26(7), 1390-1395 (2015-05-01)
Although commonly used, early initiation of salvage androgen deprivation therapy (ADT) has not been proven to enhance survival. We evaluated whether prostate-specific antigen (PSA) anxiety or health literacy are associated with use of early salvage ADT among men with recurrent
Bipolar Conductance Switching of Single Anthradithiophene Molecules
Borca et al
ACS Nano, 9, 12506-12506 (2015)
Rashmi Gaur et al.
Scientific reports, 5, 13387-13387 (2015-08-26)
This study presents genome-wide discovery of SNPs through next generation sequencing of the genome of Cicer reticulatum. Mapping of the C. reticulatum sequenced reads onto the draft genome assembly of C. arietinum (desi chickpea) resulted in identification of 842,104 genomic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service