Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L512109

Sigma-Aldrich

TBADN

AldrichCPR

Synonym(s):

2-tert-Butyl-9,10-di(naphth-2-yl)anthracene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H30
CAS Number:
Molecular Weight:
486.64
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

SMILES string

CC(C)(C)C1=CC2=C(C3=CC=C(C=CC=C4)C4=C3)C5=CC=CC=C5C(C6=CC=C(C=CC=C7)C7=C6)=C2C=C1

InChI

1S/C38H30/c1-38(2,3)31-20-21-34-35(24-31)37(30-19-17-26-11-5-7-13-28(26)23-30)33-15-9-8-14-32(33)36(34)29-18-16-25-10-4-6-12-27(25)22-29/h4-24H,1-3H3

InChI key

OBAJPWYDYFEBTF-UHFFFAOYSA-N

Application

2-tert-Butyl-9,10-di(naphth-2-yl)anthracene (TBADN) is used as a blue light-emitting material in organic light-emitting diode (OLED) [1]

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anthracene derivative for a non-doped blue-emitting organic electroluminescence device with both excellent color purity and high efficiency
Tao et al.
Chemical Physics Letters, 397, 1-1 (2004)
Yanqiu Liu et al.
Colloids and surfaces. B, Biointerfaces, 159, 596-604 (2017-09-01)
Surface properties of nanoparticles (NPs) have a huge influence on their biological activities. In this work, we report to use mesoporous silica nanoshell surface to regulate the cellular internalization rate and intracellular fate of fluorescent organic NPs for highly improved

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service