Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L380045

Sigma-Aldrich

DCI Activator

configured for ABI, 0.25M

Synonym(s):

DCI activator solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116105

type

for DNA synthesis

product line

Proligo Reagents

packaging

pkg of 6 × 450 mL L380045-06

concentration

29-32 g/L

impurities

≤30 ppm water content (Karl Fischer)
≤30 ppm water content (Karl Fischer)

color

clear Appearance of Solution
clear

compatibility

configured for ABI

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

41.0 °F

Flash Point(C)

5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service