Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

JKA5017

Sigma-Aldrich

NH2-PEG-COOH

average MN 2,000

Synonym(s):

Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51171641
NACRES:
NA.23

Product Name

NH2-PEG2K-COOH,

form

solid

storage temp.

−20°C

Application

Activated PEG derivatives can be used to modify peptides, proteins, or in other bioconjugation applications. PEGylated materials have found broad use in drug delivery systems, virology, and immunology, as the incorporation of PEG improves pharmacological properties such as increased water solubility, enhanced resistance to degradation (protein hydrolysis), increased circulation half-life, and reduced antigenicity. In addition to PEGylation, activated PEG derivatives can also be used to form networks for tissue engineering or drug delivery applications, depending on the architecture and reactivity.

Legal Information

Product of JenKem Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service