Skip to Content
MilliporeSigma

Skip To

JKA3003

mPEG20K

average Mn 20,000, Succinimidyl Carboxymethyl Ester, amine reactive

greener alternative

Greener Alternative

Synonym(s):

mPEG-Succinimidyl Carboxymethyl Ester, Polyethylene glycol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
(CH3O(CH2CH2O)nCH2COOC4H4O2N
UNSPSC Code:
12162002
NACRES:
NA.23
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

mPEG20K-Succinimidyl Carboxymethyl Ester, average Mn 20,000

form

solid

mol wt

average Mn 20,000

reaction suitability

reagent type: chemical modification reagent
reactivity: amine reactive

greener alternative product characteristics

Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

polymer architecture

shape: linear
functionality: monofunctional

storage temp.

−20°C

General description

Methoxy PEG NHS Ester (SCM PEG). Amine reactive PEG towards the amino groups of lysine(s) on proteins or other biologics; reaction occurs at room temperature in <1hr at pH 7-8. Stable linker between PEG and NHS ester.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. Polyethylene glycol (PEG) is an eco-friendly, biodegradable polymer widely used in pharmaceuticals and cosmetics. Its non-toxic nature and versatility make it a sustainable choice, derived from renewable resources, contributing to greener product formulations. Click here for more information.

Application

Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization

Legal Information

Product of JenKem Technology

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
JKA3016JKA7038JKA3033
description

average Mn 20,000

description

average Mn 10,000

description

average Mn 20,000

description

average Mn 10,000

form

solid

form

solid

form

solid

form

solid

polymer architecture

shape: linear
functionality: monofunctional

polymer architecture

shape: linear
functionality: monofunctional

polymer architecture

shape: 4-arm
functionality: homofunctional

polymer architecture

shape: linear
functionality: monofunctional

reaction suitability

reagent type: chemical modification reagent
reactivity: amine reactive

reaction suitability

reagent type: chemical modification reagent
reactivity: amine reactive

reaction suitability

reactivity: amine reactive

reaction suitability

reagent type: chemical modification reagent
reactivity: N-terminal amine reactive

mol wt

average Mn 20,000

mol wt

average Mn 10,000

mol wt

average Mn 20,000

mol wt

average Mn 10,000

storage temp.

−20°C

storage temp.

-

storage temp.

−20°C

storage temp.

−20°C

greener alternative product characteristics

Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

greener alternative product characteristics

Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

greener alternative product characteristics

-

greener alternative product characteristics

-


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters