Key Documents
HAM59000-10
Hamilton® Microlab® 600 instrument syringe
model, 1702 BFP, volume 25 μL
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
material
Gastight Borosilicate glass/PTFE syringe
feature
model 1702 BFP
manufacturer/tradename
Hamilton 59000-10
parameter
10-115 °C temp. range (50-239 °F)
1000 psig pressure
I.D. × O.D.
0.729 mm × 7.75 mm
volume
25 μL
Related Categories
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Microlab is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)