Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H52201

Sigma-Aldrich

4-Hydroxy-4-phenylpiperidine

99%

Synonym(s):

4-Phenyl-4-piperidinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15NO
CAS Number:
Molecular Weight:
177.24
Beilstein:
141577
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

157-161 °C (lit.)

SMILES string

OC1(CCNCC1)c2ccccc2

InChI

1S/C11H15NO/c13-11(6-8-12-9-7-11)10-4-2-1-3-5-10/h1-5,12-13H,6-9H2

InChI key

KQKFQBTWXOGINC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for synthesis of:
  • Diaminotriazine hNav1.7 inhibitors
  • Selective σ−ligands
  • Glycogen synthase kinase-3β−inhibitors
  • Diphenylbutylpiperidines as cell autophagy inducers
  • Malonyl-coA decarboxylase inhibitors with antiobesity and antidiabetic activities
  • Selective dopamine D2 receptor antagonists

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service