Skip to Content
MilliporeSigma
All Photos(4)

Documents

H11602

Sigma-Aldrich

Hexamethylphosphoramide

99%

Synonym(s):

HMPA, Hexamethylphosphoric acid triamide, Tris(dimethylamino)phosphine oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2N]3PO
CAS Number:
Molecular Weight:
179.20
Beilstein:
1099903
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.22

vapor density

6.18 (vs air)

Quality Level

vapor pressure

0.07 mmHg ( 25 °C)

Assay

99%

refractive index

n20/D 1.459 (lit.)

bp

230-232 °C/740 mmHg (lit.)

mp

7 °C (lit.)

density

1.03 g/mL at 25 °C (lit.)

SMILES string

CN(C)P(=O)(N(C)C)N(C)C

InChI

1S/C6H18N3OP/c1-7(2)11(10,8(3)4)9(5)6/h1-6H3

InChI key

GNOIPBMMFNIUFM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hexamethylphosphoramide is a polar aprotic solvent.

Application

Hexamethylphosphoramide can be used as a solvent:
  • For the conversion of carboxylate salt into esters by reacting with alkyl halides.
  • To synthesize aryl thioethers via nucleophilic displacement of unactivated aryl halides by thiolate ions.
  • To synthesize polyazomethines (PAZs) from dialdehyde and diamine via high-temperature solution polycondensation.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Dam. 1 - Muta. 1B - Skin Corr. 1C

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

291.2 °F - closed cup

Flash Point(C)

144 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optical and structural study of thin film of polyazomethine with triphenylamine unit prepared via spin-coating method.
Palewicz M, et al.
Polymer Bull., 66(1), 65-76 (2011)
Controlled/?Living? Reverse Atom Transfer Radical Polymerization of a Monocyclic Olefin, Methyl 1-Cyclobutenecarboxylate.
Chen X P, et al.
Macromolecules, 35(11), 4277-4281 (2002)
Hexamethylphosphoramide
Normant H.
Angewandte Chemie (International Edition in English), 6(12), 1046-1067 (1967)
Nucleophilic aromatic substitution reactions of unactivated aryl halides with thiolate ions in hexamethylphosphoramide.
Cogolli P, et al.
The Journal of Organic Chemistry, 44(15), 2642-2646 (1979)
Knud Nairz et al.
Genome biology, 5(10), R83-R83 (2004-10-06)
We report the use of the cross-linking drug hexamethylphosphoramide (HMPA), which introduces small deletions, as a mutagen suitable for reverse genetics in the model organism Drosophila melanogaster. A compatible mutation-detection method based on resolution of PCR fragment-length polymorphisms on standard

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service