Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF98079777

Gallium-indium-tin alloy, Ga68.5In21.5Sn10

250g lump, 100mm max. lump size, liquid at rtp

Synonym(s):

GA056100

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101711
NACRES:
NA.23

form

lumps (liquid at rtp)

manufacturer/tradename

Goodfellow 980-797-77

weight

250 g

max. lump size

100 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Application


  • Use of Valproic Acid-D₆ in pharmacokinetics: Study describes the use of liquid chromatography-tandem mass spectrometry (LC-MS/MS) for the determination of valproic acid and its six metabolites in human serum, highlighting the role of Valproic Acid-D₆ as an internal standard in bioanalytical methods. This application is essential for investigating drug interactions, particularly with carbapenems in epileptic patients (Yang et al., 2023).

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service