Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF92900875

Scandium

lump, 25.4 mm max. lump size, weight 5 g, purity 96+%

Synonym(s):

SC006100

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Sc
CAS Number:
Molecular Weight:
44.96
MDL number:
UNSPSC Code:
12141613
PubChem Substance ID:
NACRES:
NA.23

Assay

96%

form

lumps

manufacturer/tradename

Goodfellow 929-008-75

resistivity

50.5 μΩ-cm, 0°C

weight

5 g

max. lump size

25.4 mm

bp

2836 °C (lit.)

mp

1540 °C (lit.)

density

2.99 g/mL at 25 °C (lit.)

SMILES string

[Sc]

InChI

1S/Sc

InChI key

SIXSYDAISGFNSX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service