Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF89164873

Antimony

foil, not light tested, 25x25mm, thickness 0.002mm, permanent polyester support, 95+%

Synonym(s):

Antimony, SB000030

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Sb
CAS Number:
Molecular Weight:
121.76
MDL number:
UNSPSC Code:
12141704
PubChem Substance ID:
NACRES:
NA.23

Assay

≥95%

form

foil

manufacturer/tradename

Goodfellow 891-648-73

resistance

41.7 μΩ-cm

bp

1635 °C (lit.)

mp

630 °C (lit.)

density

6.69 g/mL at 25 °C (lit.)

SMILES string

[Sb]

InChI

1S/Sb

InChI key

WATWJIUSRGPENY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Edward R T Tiekink
Critical reviews in oncology/hematology, 42(3), 217-224 (2002-06-07)
The main group elements antimony and bismuth are used clinically, primarily for the treatment of Leishmaniasis (antimony) and ulcers (bismuth). Despite their medicinal efficacy, the exploration of the anti-cancer potential of antimony and bismuth compounds is not as well developed
Khatima Aït-Oudhia et al.
Parasitology research, 109(5), 1225-1232 (2011-07-30)
Leishmania is the causative agent of various forms of leishmaniasis, a significant cause of morbidity and mortality. The clinical manifestations of the disease range from self-healing cutaneous and mucocutaneous skin ulcers to a fatal visceral form named visceral leishmaniasis or
Ricardo Khouri et al.
The Journal of infectious diseases, 210(2), 306-310 (2014-02-11)
We show that increased plasma superoxide dismutase 1 (SOD1) levels are statistically significant predictors of the failure of pentavalent antimony treatment for cutaneous leishmaniasis caused by Leishmania braziliensis. In Leishmania amazonensis-infected patients, host SOD1 levels can be used to discriminate
Pankaj Sharma et al.
Acta pharmacologica Sinica, 29(8), 881-890 (2008-07-31)
Antimony, a natural element that has been used as a drug for over more than 100 years, has remarkable therapeutic efficacy in patients with acute promyelocytic leukemia. This review focuses on recent advances in developing antimony anticancer agents with an
M J Nash et al.
Journal of environmental monitoring : JEM, 2(2), 97-109 (2001-03-17)
Methodologies for the environmental analysis of total antimony and aqueous chemical speciation are critically reviewed, including preparation techniques for aqueous and solid matrices and the determination of solid state partitioning and recommendations are given for future research directions. Concentrations of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service