Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF87470253

Erbium

wire reel, 100mm, diameter 1.0mm, hard, 99.9%

Synonym(s):

Erbium, ER005110

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Er
CAS Number:
Molecular Weight:
167.26
MDL number:
UNSPSC Code:
12141603
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.9%

form

wire

manufacturer/tradename

Goodfellow 874-702-53

resistivity

86 μΩ-cm, 20°C

bp

2868 °C (lit.)

mp

1529 °C (lit.)

density

9.062 g/mL at 25 °C (lit.)

SMILES string

[Er]

InChI

1S/Er

InChI key

UYAHIZSMUZPPFV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Fleming
Journal of cutaneous laser therapy, 1(1), 15-21 (2001-05-22)
Carbon dioxide laser resurfacing has been a valuable procedure for facial skin rejuvenation since the early 1990s, largely replacing medium and deep chemical peels and dermabrasion. The introduction of the erbium:YAG laser for resurfacing has caused confusion about its role.
G N Marcells et al.
The Journal of otolaryngology, 29(2), 78-82 (2000-05-20)
There are two lasers for resurfacing. The erbium:YAG laser and the CO2 laser both have different physical and physiologic differences when striking the skin. Therefore, each laser gives a different result. This article reviews the use of the Sharplan Silktouch
D M Clarkson
Dental update, 28(6), 298-302 (2001-08-31)
This article reviews aspects of the probable mechanisms used by erbium dental lasers for cutting dentine and enamel, describes key issues of the risk of temperature elevation and speed of cutting relative to conventional techniques and looks at issues concerned
R Kaufmann
Clinical and experimental dermatology, 26(7), 631-636 (2001-11-07)
Laser resurfacing procedures in actinic skin damage and the treatment of disorders associated with skin ageing are becoming more popular. For both purposes Erbium:YAG laser-technology enables us to perform a highly precise skin ablative work, which can be most helpful
Nathalie Fournier et al.
Dermatologic surgery : official publication for American Society for Dermatologic Surgery [et al.], 31(9 Pt 2), 1227-1235 (2005-09-24)
Nonablative remodeling has recently been proposed as a new antiaging treatment with no downtime. This technique could greatly benefit the increasing numbers of patients who are looking for new and efficient techniques, with a minimum of discomfort. AND METHODS. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service