Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF86561354

Bismuth

foil, not light tested, 50x50mm, thickness 0.025mm, permanent polyester support, 99.97%

Synonym(s):

Bismuth, BI000160

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Bi
CAS Number:
Molecular Weight:
208.98
MDL number:
UNSPSC Code:
12141706
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.1 mmHg ( 20 °C)

Assay

≥99.97%

form

foil

manufacturer/tradename

Goodfellow 865-613-54

resistivity

129 μΩ-cm, 20°C

bp

1560 °C (lit.)

mp

271 °C (lit.)

density

9.8 g/mL at 25 °C (lit.)

SMILES string

[Bi]

InChI

1S/Bi

InChI key

JCXGWMGPZLAOME-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Javier P Gisbert et al.
Helicobacter, 10(3), 157-171 (2005-05-21)
To systematically review the Helicobacter pylori eradication efficacy with ranitidine bismuth citrate (RBC) and two antibiotics, and to conduct a meta-analysis of randomized clinical trials comparing the efficacy of proton pump inhibitor (PPI) vs. RBC with two antibiotics for 1
Montserrat Filella
Journal of environmental monitoring : JEM, 12(1), 90-109 (2010-01-19)
Like all elements of the periodic table, bismuth is ubiquitously distributed throughout the environment as a result of natural processes and human activities. It is present as Bi(III) in environmental, biological and geochemical samples. Although bismuth and its compounds are
Emma Aneheim et al.
Scientific reports, 5, 12025-12025 (2015-07-15)
To facilitate multicentre clinical studies on targeted alpha therapy, it is necessary to develop an automated, on-site procedure for conjugating rare, short-lived, alpha-emitting radionuclides to biomolecules. Astatine-211 is one of the few alpha-emitting nuclides with appropriate chemical and physical properties
Jorge A R Salvador et al.
Future medicinal chemistry, 4(11), 1495-1523 (2012-08-04)
In recent years, the chemical potential of bismuth and bismuth compounds has been actively exploited. Bismuth salts are known for their low toxicity, making them potential valuable reagents for large-scale synthesis, which becomes more obvious when dealing with products such
Syntheses of biodegradable and biocompatible polymers by means of bismuth catalysts.
Hans R Kricheldorf
Chemical reviews, 109(11), 5579-5594 (2009-09-26)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service