Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF82063032

Iron/nickel

powder, 150 max. part. size (micron), Fe58%/Ni42%, weight 200 g, condition alloy pre-cursor

Synonym(s):

FE136010

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11171604
NACRES:
NA.23

form

powder

manufacturer/tradename

Goodfellow 820-630-32

weight

200 g

max. part. size

150 (micron)

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service