Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF71822381

Aluminum-manganese alloy, Al99.35Mn0.65

100g lump, 6mm max. lump size, granular

Synonym(s):

AD086100

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352311
NACRES:
NA.23

form

lumps (granular)

manufacturer/tradename

Goodfellow 718-223-81

weight

100 g

max. lump size

6 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service