Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF70782240

Stainless Steel - AISI 316 alloy, FeCr18Ni10Mo3

mesh, 300 x 300mm, 0.103mm nominal aperture, 0.066mm wire diameter, 150 x 150 wires/inch, 0.37 open area, plain weave mesh type

Synonym(s):

FE248705

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11171600
NACRES:
NA.23

description

150 x 150 (wires/inch)

form

(mesh)

manufacturer/tradename

Goodfellow 707-822-40

diam. × nominal capacity

0.066 mm × 0.103 mm , plain weave mesh

size × opening

300 mm × 300 mm × 0.37

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service