Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF65553598

Polycarbonate

(PC), granule, 3 mm nominal granule size, weight 100 g, additives heat stabilizer only

Synonym(s):

CT306310

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352100
NACRES:
NA.23

form

granular

manufacturer/tradename

Goodfellow 655-535-98

weight

100 g

nominal granule size

3 mm

General description

For updated SDS information please visit www.goodfellow.com

Application

  • Bio-based polycarbonate as synthetic toolbox: This research explores the tuning of aliphatic polycarbonate properties, achieving features like pH-dependent water solubility and introducing a synthetic route to new polycarbonate types (O Hauenstein, S Agarwal, A Greiner, 2016).
  • Short carbon fiber reinforced polycarbonate composites: Analysis of the effects of sizing material type and level on the mechanical, electrical, and morphological properties of carbon fiber reinforced polycarbonate (C Ozkan, NG Karsli, A Aytac, V Deniz, 2014).
  • Insight into chain scission and release profiles from photodegradation of polycarbonate microplastics: This paper investigates the degradation and environmental impact of Bisphenol A polycarbonate under exposure to light, presenting a significant environmental concern (Y Shi, P Liu, X Wu, H Shi, H Huang, H Wang, S Gao, 2021).
  • Reprocessable acid-degradable polycarbonate vitrimers: Development of hydroxyl-functionalized polycarbonate networks that undergo transcarbonation exchange reactions, enabling recycling at high temperatures (RL Snyder, DJ Fortman, GX De Hoe, 2018).

Legal Information

Product of Goodfellow

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Protocols

Selenium's roles range from essential dietary element to semiconductor in advanced applications like xerography.

Selenium's roles range from essential dietary element to semiconductor in advanced applications like xerography.

Selenium's roles range from essential dietary element to semiconductor in advanced applications like xerography.

Selenium's roles range from essential dietary element to semiconductor in advanced applications like xerography.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service